missing translation for 'onlineSavingsMsg'
Learn More

Rigosertib, MedChemExpress

Product Code. 16484938
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484938 10 mg 10mg
16465716 100 mg 100mg
16451187 50 mg 50mg
16407687 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484938 Supplier Medchem Express Supplier No. HY12037A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Rigosertib (ON-01910) is a multi-kinase inhibitor and a selective anti-cancer agent, which induces apoptosis by inhibition the PI3 kinase/Akt pathway, promots the phosphorylation of histone H2AX and induces G2/M arrest in cell cycle. Rigosertib is a selective and non-ATP-competitive inhibitor of PLK1 with an IC50 of 9 nM.

Rigosertib (ON-01910) is a multi-kinase inhibitor and a selective anti-cancer agent, which induces apoptosis by inhibition the PI3 kinase/Akt pathway, promots the phosphorylation of histone H2AX and induces G2/M arrest in cell cycle. Rigosertib is a selective and non-ATP-competitive inhibitor of PLK1 with an IC50 of 9 nM.

Specifications

Chemical Name or Material Rigosertib
CAS 592542-59-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H25NO8S
Quantity 10 mg
Synonym ON-01910
Solubility Information DMSO : 75 mg/mL (166.12 mM; Need ultrasonic)
SMILES COC1=CC=C(C=C1NCC(O)=O)CS(/C=C/C2=C(C=C(C=C2OC)OC)OC)(=O)=O.[(E)]
Molecular Weight (g/mol) 451.49
Formula Weight 451.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.