missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIG-I Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£188.00 - £375.00
Specifications
| Antigen | RIG-I |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18312437
|
Novus Biologicals
NBP3-03011-20UL |
20 μL |
£188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18364956
|
Novus Biologicals
NBP3-03011-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RIG-I Polyclonal specifically detects RIG-I in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| RIG-I | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIG-IDKFZp434J1111, RNA helicase RIG-I | |
| A synthetic peptide corresponding to a sequence within amino acids 830 to the C-terminus of human RIG-I (NP_055129.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS with 50% glycerol, pH7.3. | |
| 23586 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title