missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIG-I Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33405-20ul
This item is not returnable.
View return policy
Description
RIG-I Monoclonal antibody specifically detects RIG-I in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RIG-I | |
| Monoclonal | |
| Western Blot 1:2000 - 1:4000, ELISA Recommended starting concentration is 1 μg/mL | |
| DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIG-IDKFZp434J1111, RNA helicase RIG-I | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 241-455 of human RIG-I (NP_055129.2).,, Sequence:, FKPRNYQLELALPAMKGKNTIICAPTGCGKTFVSLLICEHHLKKFPQGQKGKVVFFANQIPVYEQQKSVFSKYFERHGYRVTGISGATAENVPVEQIVENNDIIILTPQILVNNLKKGTIPSLSIFTLMIFDECHNTSKQHPYNMIMFNYLDQKLGGSSGPLPQVIGLTASVGVGDAKNTDEALDYICKLCASLDASVIATVKHNLEELEQVVYK | |
| 20 μL | |
| Phospho Specific | |
| 23586 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction