missing translation for 'onlineSavingsMsg'
Learn More

RIG-I Antibody, DyLight 594, Novus Biologicals™

Product Code. 18418245
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418245 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418245 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP227419DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RIG-I Polyclonal antibody specifically detects RIG-I in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIG-I
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIG-IDKFZp434J1111, RNA helicase RIG-I
Host Species Rabbit
Immunogen A portion of amino acid 200-250 of human RIG-I was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 23586
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.