Learn More
RIG-1/DDX58 Mouse anti-Human, Unconjugated, Clone: 3B10F5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67556-1-IG-20UL
Description
The RIG-1/DDX58 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RIG-1/DDX58. This antibody recognizes human, mouse, rat antigen. The RIG-1/DDX58 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| RIG-1/DDX58 | |
| Monoclonal | |
| 940 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 3B10F5 | |
| Unconjugated | |
| 20 μL | |
| 23586 | |
| IgG1 |