missing translation for 'onlineSavingsMsg'
Learn More

Rifampin, MP Biomedicals™

Product Code. 17680917 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17680917 250 mg 250mg
17670917 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17680917 Supplier MP Biomedicals Supplier No. 02195490.2
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibits DNA-dependent bacterial RNA polymerase. Rifampin, MP Biomedicals is a bactericidal antibiotic drug of the rifamycin group that does not affect mammalian RNA polymerase. It is used to treat Tuberculosis and Tuberculosis-related mycobacterial infec

Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase. It inhibits the assembly of DNA and protein into mature virus particles.

  • Treats Tuberculosis and Tuberculosis-related mycobacterial infections
  • Widely used as an antipruritic agent in the autoimmune cholestatic liver disease, primary biliary cirrhosis (PBC)
  • It has been shown to cause hepatitis

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rifamycin AMP, Rifampicin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampicin
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Physical Form Powder
Absorbance λ max (methanol): 237–239, 334–338, 475–479nm
Melting Point 183°C to 188°C (decomposition)(literature)
Extinction Coefficient EmM (max absorbance, phosphate buffer, pH 7.38): 33.20 (237nm); 32.10 (255nm); 27.00 (334nm);15.40 (475nm) (Lit.)
Color Orange
Quantity 250 mg
Optical Rotation [α]D25°=+10.6° (c=0.5% in CDCl3)(Lit.)
Solubility Information Soluble in chloroform (349mg/mL, 25°C), DMSO (∽100mg/mL), ethyl acetate (108mg/mL, 25°C), methanol (16mg/mL, 25°C) tetrahydrofuran.
Formula Weight 822.96
Grade Not Available
compliance-icons
Product Identifier
  • Rifampin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Rifampin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.