missing translation for 'onlineSavingsMsg'
Learn More

Rifampin, MP Biomedicals™

Product Code. 17670917 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
This item is not returnable. View return policy

Product Code. 17670917

Brand: MP Biomedicals 02195490.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits DNA-dependent bacterial RNA polymerase. Rifampin, MP Biomedicals is a bactericidal antibiotic drug of the rifamycin group that does not affect mammalian RNA polymerase. It is used to treat Tuberculosis and Tuberculosis-related mycobacterial infec

Mode of Action: Inhibits initiation of RNA synthesis by binding to β-subunit of RNA polymerase. It inhibits the assembly of DNA and protein into mature virus particles.

  • Treats Tuberculosis and Tuberculosis-related mycobacterial infections
  • Widely used as an antipruritic agent in the autoimmune cholestatic liver disease, primary biliary cirrhosis (PBC)
  • It has been shown to cause hepatitis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rifamycin AMP, Rifampicin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampicin
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Physical Form Powder
Absorbance λ max (methanol): 237–239, 334–338, 475–479nm
Melting Point 183°C to 188°C (decomposition)(literature)
Extinction Coefficient EmM (max absorbance, phosphate buffer, pH 7.38): 33.20 (237nm); 32.10 (255nm); 27.00 (334nm);15.40 (475nm) (Lit.)
Quantity 100 mg
Optical Rotation [α]D25°=+10.6° (c=0.5% in CDCl3)(Lit.)
Solubility Information Soluble in chloroform (349mg/mL, 25°C), DMSO (∽100mg/mL), ethyl acetate (108mg/mL, 25°C), methanol (16mg/mL, 25°C) tetrahydrofuran.
Formula Weight 822.96
compliance-icons
Product Identifier
  • Rifampin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Rifampin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.