missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rifampin, Molecular Biology Reagent

Produktkod. p-7041957
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Förpackningsstorlek
5g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15473539

missing translation for 'mfr': Thermo Scientific Chemicals J60836.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rifampin (rifampicin), CAS # 13292-46-1, is a synthetic rifamycin antibiotic that works by inhibiting RNA and DNA synthesis and exhibits activity against the tuberculosis microbe. | CAS: 13292-46-1 | C43H58N4O12 | 822.953 g/mol

Antibiotic effective against mycobacteria. Can also inhibit DNA polymeraseRifampin is an antibiotic used in the treatment of tuberculosis and its related microbacterial infections. It is involved in the inhibition of DNA polymerase and RNA polymerase. As an antipruritic agent, it is used in the autoimmune cholestatic liver disease and primary biliary cirrhosis (PBC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Rifampin is a rifamycin class antibiotic synthesized to resemble the compound isolated from Amycolatopsis rifamycinica

Applications

  • Rifampin can be used as a bacteriostatic that prevents RNA and DNA synthesis, particularly in mycobacteria
  • It may also act as an antineoplastic, a neuroprotective, and inhibitor of angiogenesis
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
MDL Number MFCD00151389
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rifampicin
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifikationer

Type Rifampicin
Physical Form Powder
Melting Point 183°C to 188°C
Color Brown/Red to Orange
Quantity 5 g
Beilstein 5723476
Merck Index 14,8216
Solubility Information Soluble in DMSO or methanolSoluble in water,ethyl acetate,chloroform,methanol,tetrahydrofuran and dimethyl sulfoxide.
Formula Weight 822.94
Grade Molecular Biology
Chemical Name or Material Rifampin
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Rifampin, Molecular Biology Grade
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Kompletterande information
  • MIXTURE LIST-Contains : Rifampicin

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.