missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rifampin, 95%

Product Code. 15333786 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15333786 25 g 25g
15515166 1 g 1g
15513101 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15333786 Supplier Thermo Scientific Chemicals Supplier No. 455620250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rifampin (rifampicin), CAS # 13292-46-1, is a synthetic rifamycin antibiotic that works by inhibiting RNA and DNA synthesis and exhibits activity against the tuberculosis microbe.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Rifampin is a rifamycin class antibiotic synthesized to resemble the compound isolated from Amycolatopsis rifamycinica

Applications

• Rifampin can be used as a bacteriostatic that prevents RNA and DNA synthesis, particularly in mycobacteria

• It may also act as an antineoplastic, a neuroprotective, and inhibitor of angiogenesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Type Rifampicin
Melting Point 183.0°C
Infrared Spectrum Authentic
Loss on Drying 2% max. (100 mg, 60°C, 4 h) (vacuum)
Quantity 25 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 822.94
Percent Purity 95%
Chemical Name or Material Rifampin
compliance-icons
Product Identifier
  • Rifampin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.