missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rifampicin, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Rifampicin |
| CAS | 13292-46-1 |
| Analyte or Component Names | Rifampicin |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C43 H58 N4 O12 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Rifampicin, (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-5,6,9,17,19-Pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-8-[[(4-methylpiperazin-1-yl)imino]methyl]-1,11-dioxo-1,2-dihydro-2,7-(epoxypentadeca[1,11,13]trienimino)naphto[2,1-b]furan-21-yl acetate, Rifamycin, 3-[[(4-methyl-1-piperazinyl)imino]methyl]-, -(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione, 5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl-1-piperazinyl)formimidoyl]-, 21-acetate (8CI), 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione, 5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,20,22-heptamethyl-8-[N-(4-methyl-1-piperazinyl)formimidoyl]-, 21-acetate (7CI), 2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan, rifamycin deriv., 3-[(4-Methyl-1-piperazinyl)iminomethyl]rifamycin SV, 5,6,9,17,19,21-Hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl-1-piperazinyl)formimidoyl]-2,7-(epoxypentadeca[1,11,13]trienimino)-naphtho[2,1-b]furan-1,11(2H)-dione 21-acetate, Abrifam, Ba 41166/E, Eremfat, Famcin, L 5103 Lepetit, NIH 10782, NSC 113926, R-Cin, R/AMP, Refampicin, Rifa, Rifacap, Rifadin, Rifadine, Rifaldazine, Rifaldin, Rifampicin, Rifampicin SV, Rifampin, Rifamycin AMP, Rifaprodin, Rifodex, Rifoldin, Rifoldine, Riforal, Rimactan, Rimactane, Rimapen, Sinerdol, Tubocin |
| InChI Formula | InChI=1S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)39(58-27(7)48)25(5)35(50)24(4)34(21)49/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13-,44-20+/t21-,23+,24+,25+,29-,34-,35+,39+,43-/m0/s1 |
| Solution Type | Neat |
| SMILES | CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C)c(\C=N\N5CCN(C)CC5)c(O)c4c3C2=O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction