Learn More
Melford Rifampicin
Description
Rifampicin is an antibiotic used for its potent antibacterial properties.
Rifampicin is an antibiotic used for its potent antibacterial properties. It belongs to the class of antibiotics known as rifamycins. Rifampicin is particularly effective against bacteria, including Mycobacterium tuberculosis, which causes tuberculosis, and other bacteria that can lead to various infections.
The mechanism of action of rifampicin involves inhibiting bacterial RNA synthesis by binding to the beta subunit of bacterial DNA-dependent RNA polymerase. This interference disrupts the transcription process, preventing the synthesis of essential RNA and proteins needed for bacterial growth and survival.
Rifampicin is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its use contributes to the identification and study of specific bacterial strains.
Specifications
Specifications
| Chemical Name or Material | Rifampicin |
| CAS | 13292-46-1 |
| Quantity | 1 g |
| Molecular Formula | C43H58N4O12 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.