missing translation for 'onlineSavingsMsg'
Learn More

Rifabutin, MedChemExpress

Product Code. 16473287
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473287 50 mg 50mg
16471826 500 mg 500mg
16447236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473287 Supplier Medchem Express Supplier No. HY1702550mg
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rifabutin (Ansamycin) is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties. Rifabutin inhibits DNA-dependent RNA polymerase.

Rifabutin (Ansamycin) is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties. Rifabutin inhibits DNA-dependent RNA polymerase.

Specifications

Chemical Name or Material Rifabutin
Physical Form Solid
CAS 72559-06-9
Color Deep Purple
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C46H62N4O11
For Use With (Application) COVID-19-anti-virus
Synonym Ansamycin LM-427
Solubility Information DMSO : 50 mg/mL (59.03 mM; Need ultrasonic)
SMILES C[C@@]1(O2)O/C=C/[C@@H]([C@H]([C@H]([C@H](C)[C@H](O)[C@H](C)[C@H]([C@H](/C=C/C=C(C(NC(C3=O)=C4NC5(CCN(CC(C)C)CC5)N=C4C6=C3C(O)=C(C)C2=C6C1=O)=O)\C)C)O)OC(C)=O)C)OC
Molecular Weight (g/mol) 847.0
Formula Weight 847.0
Percent Purity 99.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.