missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIF1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30119-1-AP-20UL
This item is not returnable.
View return policy
Description
The RIF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RIF1. This antibody recognizes human, mouse antigen. The RIF1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| RIF1 | |
| Polyclonal | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| 300 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 600 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55183 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur