missing translation for 'onlineSavingsMsg'
Learn More

Ridinilazole, MedChemExpress

Product Code. 16450576
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16450576

Brand: Medchem Express HY16753

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ridinilazole is a novel antibacterial with MICs range of 0.06-0.25 μg/mL (MIC90=8 μg/mL) against C. difficile.

Ridinilazole is a novel antibacterial with MICs range of 0.06-0.25 μg/mL (MIC90=8 μg/mL) against C. difficile.

Specifications

Chemical Name or Material Ridinilazole
Physical Form Solid
CAS 308362-25-6
Color Yellow
Quantity 20 mg
Purity Grade Notes Research
Molecular Formula C24H16N6
For Use With (Application) COVID-19-immunoregulation
Synonym SMT19969
Solubility Information DMSO : ≥ 60 mg/mL (154.47 mM)
SMILES C1(C2=CC=NC=C2)=NC3=CC(C4=CC=C5C(N=C(C6=CC=NC=C6)N5)=C4)=CC=C3N1
Molecular Weight (g/mol) 388.42
Formula Weight 388.42
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.