missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RICTOR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27248-1-AP-20UL
This item is not returnable.
View return policy
Description
RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.Specifications
| RICTOR | |
| Polyclonal | |
| Unconjugated | |
| RICTOR | |
| AVO3 homolog, hAVO3, KIAA1999, mAVO3, RICTOR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 253260, 78757 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6QI06, Q6R327 | |
| RICTOR | |
| RICTOR Fusion Protein Ag25649 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction