missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rictor Antibody (1A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32903
This item is not returnable.
View return policy
Description
Rictor Monoclonal antibody specifically detects Rictor in Human,Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Rictor | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:500 | |
| AVO3, AVO3 homolog, hAVO3, KIAA1999DKFZp686B11164, mAVO3, MGC39830, PIA, pianissimo, rapamycin-insensitive companion of mTOR, RPTOR independent companion of MTOR, complex 2, TORC2-specific protein AVO3 | |
| Recombinant protein within human Rictor aa 1459-1708/ 1708. (Uniprot: Q6R327) | |
| 100 μL | |
| Apoptosis, Autophagy, Cancer, DNA Repair, Hypoxia, mTOR Pathway | |
| 253260 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| 1A5 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction