missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ricinus Communis (Castor Bean) Agglutinin I (RCA I), fluorescein (FITC)

Product Code. 17126642
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17126642 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17126642 Supplier Invitrogen™ Supplier No. L32477

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bright-green RCA-fluorophor conjugate

  • Ricinus Communis (Castor Bean) Agglutinin I (RCA I, RCA120), fluorescein (FITC), is a bright-green RCA-fluorophor conjugate.
  • RCA I is a tetramer composed of two α– and two β–chains. RCA I blocks protein synthesis and is toxic to cultured cells and somewhat toxic to mice.
  • RCA I has been used to distinguish tumor cells from normal cells in tissue. RCA I has also been used to distinguish microglia in human nervous tissues, where RCA I does not react with astrocytes, oligodendrocytes, and neurons.
  • RCA I does react with blood cells and endothelial cells, but those cells can be easily distinguished from microglia.
  • Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: galactose, lactose
  • Inhibiting/eluting sugar: 200 mM galactose or lactose
  • Blood group specificity: non-specific
  • Unconjugated lectin molecular weight: 120 KDa
  • Immunofluorescence working concentration: 5–20 μg/mL
  • RCA I-FITC is provided at a 5 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+
  • Caution in use of this lectin: It has been found to have moderate toxicity in some animal models, and toxicity to humans has not been determined. Please note that the related protein RCA II (RCA 60) is highly toxic to humans.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Fluorescein
Quantity 5 mg
Source Castor bean
Content And Storage When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept.

Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Family Lectins
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.