missing translation for 'onlineSavingsMsg'
Learn More

Ricinic Acid Methyl Ester, TRC

Product Code. 30427039
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427039 100 mg 100mg
30412749 1 g -
30409425 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427039 Supplier Toronto Research Chemicals Supplier No. TRCR418050100MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-24-2
Molecular Formula C19 H36 O3
Molecular Weight (g/mol) 312.49
Synonym 9-Octadecenoic acid, 12-hydroxy-, methyl ester, (9Z,12R)-, 9-Octadecenoic acid, 12-hydroxy-, methyl ester, [R-(Z)]-, Ricinoleic acid, methyl ester (6CI,8CI), Edenor Me-Ri 90, Flexricin P 1, K-PON 180, K-PON 180HP, Methyl (R)-ricinoleate, Methyl 12-D-hydroxy-9-cis-octadecenoate, Methyl cis-(R)-12-hydroxyoctadec-9-enoate, Methyl ricinate, Methyl ricinoleate, NSC 1254, Ricinic acid methyl ester, Ricinoleic methyl ester, cis-Ricinoleic acid methyl ester
IUPAC Name methyl (Z,12R)-12-hydroxyoctadec-9-enoate
SMILES CCCCCC[C@@H](O)C\C=C/CCCCCCCC(=O)OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H36O3/c1-3-4-5-12-15-18(20)16-13-10-8-6-7-9-11-14-17-19(21)22-2/h10,13,18,20H,3-9,11-12,14-17H2,1-2H3/b13-10-/t18-/m1/s1
Formula Weight 312.2664
Chemical Name or Material Ricinoleic acid methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.