Learn More
Epredia™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes, White
Ensure positive identification from grossing to filing with the three-sided writing surface of Epredia™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
Brand: Epredia™ 58930
Code : 50
Additional Details : Weight : 2.26799kg
Description
- Hinge design ensures specimen safety during processing and easy removal of cover while embedding
- Base is compatible with metal lids and fits all popular size base molds (metal and disposable plastic)
- Cassettes are conveniently available assembled or unassembled to facilitate usage in a labeling system or with metal lids
- Solvent- and decalcifier-resistant and compatible with microwave techniques
- Different colors allow for easy specimen coding
Specifications
Small Square Pore | |
Hinged | |
White | |
Biopsy Cassette |