missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS), Schiff Reagent

Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Richard-Allan Scientific™ Periodic Acid-Schiff (PAS) Solution

Brand:  Epredia™ 88017

108.19 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.



Code : A0

Additional Details : CAS Number : 7647-01-0 Weight : 0.50000kg

 View more versions of this product

Product Code. 12503647

  • £119.00 / 500mL
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

  • Glycogen, mucin, fungal walls and some basement membranes stain rose to magenta
  • Nuclei stain blue
  • Kit includes: Hematoxylin I, periodic acid solution (0.5%), and Schiff reagent
Specifications

Specifications

Schiff Reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers