Learn More
Epredia™ Richard-Allan Scientific™ Modified Wright Stain Pack
Product Code. 10539408 Shop All Epredia Products
Click to view available options
Type:
Modified Wright Stain Pack
Unit Size:
Pack of 5
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10539408
Brand: Epredia™ 3000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Achieve optimum performance on Hema-Tek™ Slide Stainers with the Epredia™ Richard-Allan Scientific™ Stain Pack.
This system consists of a modified polychrome methylene blue-eosin stain (similar to Wright or Giemsa) and based on the original stain proposed by Romanowsky*.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 Pk.
Type Modified Wright Stain Pack