missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Mark-It™ Tissue Marking Dyes

Product Code. p-4532404 Shop All Epredia Products
Click to view available options
Product Type:
Tissue Marking Dye
Color:
Black
Green
Orange
Red
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12698066

Brand: Epredia™ 3120125

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For a variety of tissue identification purposes use Epredia™ Mark-It™ Tissue Marking Dyes.

Features:
  • Permanent dyes that maintain their original brilliance during routine tissue processing cycles
  • Can be used on fresh or formalin-fixed tissues

Uses include:

  • Marking surgical margins
  • Identifying serial segments of large tissues to ensure proper orientation and embedding
  • Maintaining proper tissue orientation for Mohs surgery specimens
  • Identification of multiple pieces of tissues processed in the same cassette

Ordering Information:

  • Available in black, blue, green, orange, red and yellow in 2 oz. bottles
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Product Type Tissue Marking Dye
Quantity 2 oz./ ea.
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H371-May cause damage to organs.
Precautionary Statement
  • P362-Take off contaminated clothing.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : carbon black, ammonium hydroxide, formaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.