Learn More
Epredia™ HistoGel™ Specimen Processing Gel
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
Brand:  Epredia™ HGSK-2050-1
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12823643
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Specifications
Specifications
Twelve 10 mL HistoGel vials, two Epredia™ Richard-Allan Scientific WonderBlock cooling blocks, dry bath incubator, and disposable pipets (500)
Specimen Processing Gel