Learn More
Epredia™ HistoGel™ Specimen Processing Gel
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
Brand:  Epredia™ HG-4000-144
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12056689
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Specifications
Specifications
Vial
Specimen Processing Gel
144 Pk.