Learn More
Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), Gomori Trichrome Stain Solution, Blue Collagen
Product Code. 12523657 Shop All Epredia Products
Product Code. 12523657
Brand: Epredia™ 88030
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Save time with Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome.
  • Used to study the diseases of connective tissue and muscle associated with fibrotic and dystrophic changes
  • Stains collagen blue, cytoplasm and muscle fibers red and the nuclei blue-to-black
TRUSTED_SUSTAINABILITY
Specifications
Type Gomori Trichrome (Blue Collagen)
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : acetic acid, disodium-4,5-dihydroxy-3-phenylazonaphthalene-2,7-disulphonate, phosphotungstic acid, C