Learn More
Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), Weigerts Iron Hematoxylin, Part B
Product Code. 12513657 Shop All Epredia Products
Product Code. 12513657
Brand: Epredia™ 88029
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Save time with Epredia™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome.
  • Used to study the diseases of connective tissue and muscle associated with fibrotic and dystrophic changes
  • Stains collagen blue, cytoplasm and muscle fibers red and the nuclei blue-to-black
TRUSTED_SUSTAINABILITY
Specifications
Type Weigert's Iron Hematoxylin- Part B
compliance-icons
Product Identifier
  • SOLUTION WGRTS IRONHEMAB 500ML
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.