Learn More
Epredia™ Dry Specimen Containers - Snap Lid
Shop All Epredia Products
Click to view available options
Capacity (Metric):
360 mL
Unit Size:
Pack of 350
Description
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
- Easy to transport trays ideal for operating rooms and hospitals
- Clear polystyrene containers have screw-top lids
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
- Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Easy to transport trays
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
- Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Stack pack reduces excess packaging
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Specifications
| Product Type | Dry Specimen Container |
| Closure Included | Yes |
| Material | Polyethylene |
| Sterility | Non-sterile |
| Capacity (Metric) | 360 mL |
| Closure Type | Snap Cap |
| Color | Opaque |
| No. per Case | 350/Cs. |