Learn More
Epredia™ Dry Specimen Containers - Snap Lid
Product Code. 12530027 Shop All Epredia Products
Click to view available options
Capacity (Metric):
5.09 L
Unit Size:
Pack of 10
Product Code. 12530027
Brand: Epredia™ 41754
Dry specimen containers constructed of clear and opaque polystyrene, polypropylene or polyethylene with a variety of lid options.
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
  • Easy to transport trays ideal for operating rooms and hospitals
  • Clear polystyrene containers have screw-top lids
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
  • Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Easy to transport trays
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
  • Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Stack pack reduces excess packaging
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dry Specimen Container
Closure Included Yes
Material Polyethylene
Sterility Non-sterile
Capacity (Metric) 5.09 L
Closure Type Snap Cap
Color Opaque
No. per Case 10/Cs.