Learn More
Epredia™ Richard-Allan Scientific™ Amyloid Stain, Sodium Hydroxide Solution with dropper
Product Code. 12503637 Shop All Epredia Products
Click to view available options
Type:
Hematoxylin
Unit Size:
500mL
Product Code. 12503637
Brand: Epredia™ 88006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Demonstrate amyloid with Epredia™ Richard-Allan Scientific™ Amyloid Stain.
  • Alkaline Congo Red solution will stain amyloid orange-to-red and display apple-green birefringence when examined with polarizing microscope
TRUSTED_SUSTAINABILITY
Specifications
Type Hematoxylin
Quantity 500mL