Learn More
Invitrogen™ RICH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121092
Description
Positive test controls include: MCF7, 293T, Mouse brain, Mouse heart, Mouse lung, Rat heart, Rat brain. The target is usually found in the following locations: Cell junction, Cell projection, Recycling endosome, dendritic spine, synapse. Immunogen sequence: MADQSAGQPS PVSLSPTPPS TPSPYGLSYP QGYSLASGQL SPAAAPPLAS PSVFTSTLSK SRPTPKPRQR PTLPPPQPPT VNLSASSPQS TEAPMLDGMS PGESMST.
GTPase-activating protein (GAP) that stimulates the GTPase activity of Rho-type GTPases. Thereby, controls Rho-type GTPases cycling between their active GTP-bound and inactive GDP-bound states. Acts as a GAP at least for CDC42 and RAC1 (PubMed:11431473). In neurons, is involved in dendritic spine formation and synaptic plasticity in a specific RAC1-GAP activity. Limits the initiation of exploratory dendritic filopodia. Recruited to actin-patches that seed filopodia, binds specifically to plasma membrane sections that are deformed inward by acto-myosin mediated contractile forces. Acts through GAP activity on RAC1 to reduce actin polymerization necessary for filopodia formation. In association with SHANK3, promotes GRIA1 exocytosis from recycling endosomes and spine morphological changes associated to long-term potentiation. [UniProt].
Specifications
| RICH2 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP44 | |
| ARHGAP44; KIAA0672; NPC-A-10; Rho GTPase activating protein 44; rho GTPase-activating protein 44; rho GTPase-activating protein RICH2; RhoGAP interacting with CIP4 homologs protein 2; Rho-type GTPase-activating protein RICH2; RICH2; RICH-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 216831, 303222, 9912 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| F1LQX4, Q17R89, Q5SSM3 | |
| ARHGAP44 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 666-772 of human ARHGAP44 (NP_055674.4). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.