missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562436
This item is not returnable.
View return policy
Description
Immunogen sequence: YCATLDGFAV VFNDGKVGFI TPVSSRFTAE QLHGVWPQDV VDGTCVAVNN KYRLMAFGCV SGSVQVYTID NSTGAMLLSH KLELTAKQYP D Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.
Specifications
| RIC1 | |
| Polyclonal | |
| Unconjugated | |
| RIC1 | |
| bA207C16.1; CIP150; connexin 43-interacting protein 150 kDa; connexin-43-interacting protein of 150 kDa; KIAA1432; protein RIC1 homolog; RAB6A GEF complex partner 1; RAB6A-GEF complex partner protein 1; RIC1; RIC1 homolog, RAB6A GEF complex partner 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4ADV7 | |
| RIC1 | |
| Recombinant protein corresponding to Human RIC1. Recombinant protein control fragment (Product #RP-92903). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction