missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ribulose-phosphate 3-epimerase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557689
This item is not returnable.
View return policy
Description
Immunogen sequence: VEPGFGGQKF MEDMMPKVHW LRTQFPSLDI EVDGGVGPDT VHKCAEAGAN MIVSGSAIMR SEDPRSVINL LRNVCSEAAQ KR Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Catalyzes the reversible epimerization of D-ribulose 5-phosphate to D-xylulose 5-phosphate. [UniProt].
Specifications
| Ribulose-phosphate 3-epimerase | |
| Polyclonal | |
| Unconjugated | |
| RPE | |
| 2810429B02Rik; 5730518J08Rik; HUSSY-17; ik:tdsubc_2c8; PE; Ribulose-5-phosphate-3-epimerase; ribulose-5-phosphate-epimerase; Ribulose-phosphate 3-epimerase; Rpe; R-PE; RPE2-1; R-phycoerythrin; wu:fa07h08; wu:fb93d11; wu:fc20b11; xx:tdsubc_2c8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6120 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96AT9 | |
| RPE | |
| Recombinant protein corresponding to Human RPE. Recombinant protein control fragment (Product #RP-96231). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction