missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosome maturation protein SBDS Antibody (1E8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22594
This item is not returnable.
View return policy
Description
Ribosome maturation protein SBDS Monoclonal specifically detects Ribosome maturation protein SBDS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Ribosome maturation protein SBDS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:3000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| FLJ10917, ribosome maturation protein SBDS, SDSCGI-97, Shwachman-Bodian-Diamond syndrome, Shwachman-Bodian-Diamond syndrome protein, SWDS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human SBDS mAb, clone 1E8, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human SBDS protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 100000000 | |
| Unconjugated | |
| NP_057122 | |
| SBDS | |
| Recombinant human SBDS (1-250aa) purified from E. coli | |
| 0.1 mL | |
| metabolism | |
| 51119 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction