missing translation for 'onlineSavingsMsg'
Learn More

Ribosomal Protein S6/RPS6 Rabbit anti-Human, Mouse, Rat, Clone: 4C7P4, Novus Biologicals™

Product Code. 18341525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341525 100 μg 100µL
18389251 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18341525 Supplier Novus Biologicals Supplier No. NBP315435100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribosomal Protein S6/RPS6
Applications Western Blot
Classification Monoclonal
Clone 4C7P4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human Ribosomal Protein S6/RPS6 (NP_001001.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, mTOR Pathway, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 6194
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.