missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal Protein S6/RPS6, p Ser240, p Ser244 Antibody, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Ribosomal Protein S6/RPS6 Polyclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Ribosomal Protein S6/RPS6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 1% BSA, 20% Glycerol (pH7) |
| Gene Alias | Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 |
| Host Species | Rabbit |
| Immunogen | Carrier-protein conjugated synthetic peptide corresponding to residues around human Ribosomal Protein S6/RPS6 (phospho Ser240/Ser244). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?