missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Ribosomal Protein S6/RPS6 Polyclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Ribosomal Protein S6/RPS6 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:20 to 1:50, Immunocytochemistry/ Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:20 to 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol |
| Gene Alias | Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: GCQKLIEVDDERKLRTFYEKRMATEVAADALGEEWKGYVVRISGGNDKQGFPMKQGVLTHGRVRLLLSKGHSCYRPRRTGERKRKSVRGCIVDANLSVLNLVIVKKGEKDIPGLTDTTVPRRLGPKRASRIRKLFNLSK |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?