missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal Protein S6/RPS6 Antibody (A6B8-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32919
This item is not returnable.
View return policy
Description
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Ribosomal Protein S6/RPS6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 | |
| Recombinant protein within human Ribosomal Protein S6/RPS6 aa 1-150. (Uniprot: P62753) | |
| 100 μL | |
| Cancer, DNA Repair, mTOR Pathway, Phospho Specific, Translation Control | |
| 6194 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| A6B8-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction