missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal Protein S6/RPS6 Antibody (522731) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5436JF635
This item is not returnable.
View return policy
Description
Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
Specifications
| Ribosomal Protein S6/RPS6 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6 | |
| E. coli-derived recombinant human RPS6, Met1-Lys249, Accession # P62753 | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 522731 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, DNA Repair, mTOR Pathway, Phospho Specific, Translation Control | |
| 6194 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction