Learn More
Ribosomal protein L4 Mouse anti-Human, Unconjugated, Clone: 3C7A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67028-1-IG-20UL
Description
The Ribosomal protein L4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Ribosomal protein L4. This antibody recognizes human, mouse, rat antigen. The Ribosomal protein L4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Ribosomal protein L4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| 3C7A8 | |
| Unconjugated | |
| 20 μL | |
| 6124 | |
| IgG1 |