missing translation for 'onlineSavingsMsg'
Learn More

Ribosomal protein L26 Antibody, PE, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP2-22233PE

Product Code. 30039333

  • £467.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ribosomal protein L26 Polyclonal antibody specifically detects Ribosomal protein L26 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ribosomal protein L26
Polyclonal
Western Blot, Immunoprecipitation (Negative)
ribosomal protein L26,60S ribosomal protein L26
The immunogen this antibody was made to, maps to a region between residue 95 and 145 of human Ribosomal Protein L26 using the numbering given in entry NP_000978.1 (GeneID 6154).
0.1 mL
Cellular Markers
6154
Store at 4C in the dark.
IgG
Western Blot
PE
PBS
Goat
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.