missing translation for 'onlineSavingsMsg'
Learn More

Ribosomal protein L26 Antibody, Novus Biologicals™

Product Code. 18189561 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18189561 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18189561 Supplier Novus Biologicals Supplier No. NBP222233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Ribosomal protein L26 Polyclonal specifically detects Ribosomal protein L26 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribosomal protein L26
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Immunoprecipitation (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_000978.1
Gene Alias ribosomal protein L26,60S ribosomal protein L26
Gene Symbols RPL26
Host Species Goat
Immunogen The immunogen this antibody was made to, maps to a region between residue 95 and 145 of human Ribosomal Protein L26 using the numbering given in entry NP_000978.1 (GeneID 6154).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 6154
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.