missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal protein L26 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-22233AF647
This item is not returnable.
View return policy
Description
Ribosomal protein L26 Polyclonal antibody specifically detects Ribosomal protein L26 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Ribosomal protein L26 | |
| Polyclonal | |
| Western Blot | |
| ribosomal protein L26,60S ribosomal protein L26 | |
| The immunogen this antibody was made to, maps to a region between residue 95 and 145 of human Ribosomal Protein L26 using the numbering given in entry NP_000978.1 (GeneID 6154). | |
| 0.1 mL | |
| Cellular Markers | |
| 6154 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction