missing translation for 'onlineSavingsMsg'
Learn More

Ribose-5-phosphate Barium Salt Hydrate, TRC

Product Code. 30409468
Change view
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409468 2 mg 2mg
30424830 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30409468 Supplier Toronto Research Chemicals Supplier No. TRCR5907032MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ribose-5-phosphate Barium Salt Hydrate
CAS 15673-79-7
Molecular Formula C5 H9 O8 P . Ba
Quantity 2 mg
InChI Formula InChI=1S/C5H11O8P.Ba/c6-3-2(1-12-14(9,10)11)13-5(8)4(3)7;/h2-8H,1H2,(H2,9,10,11);/q;+2/p-2/t2-,3-,4-,5?;/m1./s1
SMILES [Ba+2].OC1O[C@H](COP(=O)([O-])[O-])[C@@H](O)[C@H]1O
IUPAC Name barium(2+);[(2R,3S,4R)-3,4,5-trihydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol) 365.421
Formula Weight 365.909
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.