missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ribophorin II Monoclonal Antibody (OTI1G9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525935
This item is not returnable.
View return policy
Description
Ribophorin II Monoclonal Antibody for Western Blot, IHC (P)
RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
Specifications
| Ribophorin II | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04844 | |
| RPN2 | |
| Full length human recombinant protein of RPN2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G9 | |
| Unconjugated | |
| RPN2 | |
| 1300012C06Rik; AV261018; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2; hypothetical protein LOC534231; oligosaccharyltransferase complex subunit (non-catalytic); RIBIIR; ribophorin 2; ribophorin II; Ribophorin-2; RPN2; Rpn-2; RPNII; RPN-II; SWP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6185 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction