missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribophorin II Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32178DL594
This item is not returnable.
View return policy
Description
Ribophorin II Polyclonal antibody specifically detects Ribophorin II in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Ribophorin II | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| RIBIIREC 2.4.1.119, ribophorin IIDolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit, Ribophorin-2, RPNII, RPN-IIdolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2, SWP1 | |
| The immunogen this antibody was made to, maps to a region between residue 350 to 400 of human Ribophorin II using the numbering given in entry NP_002942.2 (GeneID 6185). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6185 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction