missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ribophorin I Recombinant Rabbit Monoclonal Antibody (24GB1090)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557690
This item is not returnable.
View return policy
Description
Ribophorin I Recombinant Monoclonal Antibody for Western Blot, Flow
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombinant Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
Specifications
| Ribophorin I | |
| Recombinant Monoclonal | |
| Unconjugated | |
| RPN1 | |
| AU018702; D6Wsu137e; Dolichyl-diphosphooligosaccharide--protein; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); OST1; QccE-21639; QtsA-18349; RBPH1; RIBI; ribophorin I; Ribophorin-1; RPN1; Rpn-1; RPN-I; unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 103963, 25596, 6184 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1090 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04843, P07153, Q91YQ5 | |
| RPN1 | |
| A synthesized peptide derived from human PRibophorin I (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction