missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribonuclease Inhibitor Antibody (1H23) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-30167
This item is not returnable.
View return policy
Description
Ribonuclease Inhibitor Monoclonal specifically detects Ribonuclease Inhibitor in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Ribonuclease Inhibitor | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Placental ribonuclease inhibitor, Placental RNase inhibitor, PRI, RAIMGC4569, ribonuclease inhibitor, ribonuclease/angiogenin inhibitor, ribonuclease/angiogenin inhibitor 1MGC18200, RNHMGC54054 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human RNH1 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunofluorescence | |
| 1H23 | |
| Unconjugated | |
| NP_002930 | |
| RNH1 | |
| Ribonuclease inhibitor 1(RNH1) (7-461aa) purified from E. coli | |
| 0.1 mL | |
| metabolism, Proteases & Other Enzymes | |
| 6050 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction