missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribonuclease A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38229-100ul
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Ribonuclease A Polyclonal antibody specifically detects Ribonuclease A in Mouse samples. It is validated for ELISA,Western Blot
Tekniske data
| Ribonuclease A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-156 of human Ribonuclease A (NP_937875.1).,, Sequence:, KESRAKKFQRQHMDSDSSPSSSSTYCNQMMRRRNMTQGRCKPVNTFVHEPLVDVQNVCFQEKVTCKNGQGNCYKSNSSMHITDCRLTNGSRYPNCAYRTSPKERHIIVACEGSPYVPVHFDASVEDST | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 6035 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion