missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribonuclease A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38229-20ul
This item is not returnable.
View return policy
Description
Ribonuclease A Polyclonal antibody specifically detects Ribonuclease A in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Ribonuclease A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 3.1.27.5, HP-RNase, MGC12408, RIB1, RIB-1, Ribonuclease 1, Ribonuclease A, ribonuclease pancreatic, ribonuclease, RNase A family, 1 (pancreatic), RNase A, RNase UpI-1, RNS1RNase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-156 of human Ribonuclease A (NP_937875.1).,, Sequence:, KESRAKKFQRQHMDSDSSPSSSSTYCNQMMRRRNMTQGRCKPVNTFVHEPLVDVQNVCFQEKVTCKNGQGNCYKSNSSMHITDCRLTNGSRYPNCAYRTSPKERHIIVACEGSPYVPVHFDASVEDST | |
| 20 μL | |
| DNA replication Transcription Translation and Splicing, Proteases & Other Enzymes | |
| 6035 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction