missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin (Vitamin B2), Dr. Ehrenstorfer

Product Code. 30322863
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322863 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322863 Supplier Dr. Ehrenstorfer Supplier No. DREC16813600100MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Vitamin B2 (Riboflavin)
CAS 83-88-5
Analyte or Component Names Vitamin B2 (Riboflavin)
Shipping Condition +20°C
Molecular Formula C17 H20 N4 O6
Certifications/Compliance ISO 17034
Synonym Riboflavin, Riboflavine (7CI), Benzo[g]pteridine, riboflavin deriv., (-)-Riboflavin, 1-Deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10(2H)-yl)-D-ribitol, 6,7-Dimethyl-9-D-ribitylisoalloxazine, 6,7-Dimethyl-9-ribitylisoalloxazine, Beflavin, Beflavine, Benzo[g]pteridine-2,4(3H,10H)-dione, 7,8-dimethyl-10-(D-ribo-2,3,4,5-tetrahydroxypentyl)-, C.I. 50900, C.I. Food Yellow 15, D-Ribitol, 1-deoxy-1-(3,4-dihydro-7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10(2H)-yl)-, E 101, E 101 (dye), Flavaxin, Flavin BB, Flaxain, Food Yellow 15, Hyre, Lactobene, Lactoflavin, Lactoflavine, NCI 0033298, NSC 33298, Ribipca, Ribocrisina, Riboderm, Ribosyn, Ribotone, Ribovel, Russupteridine yellow III, San Yellow B, Vitaflavine, Vitamin B2, Vitamin G, Vitasan B2
InChI Formula InChI=1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1
SMILES Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)CO)c2cc1C
IUPAC Name 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione
Molecular Weight (g/mol) 376.36
Quantity 100 mg
Formula Weight 376.1383 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.